missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-DL-phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™

Catalog Number 15498196
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14173-40-1
C10H13Cl2NO2
250.119
MFCD00012530
GCBCWTWQAFLKJG-UHFFFAOYSA-N
4-chloro-dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, h-dl-phe 4-cl-ome.hcl, dl-4-chlorophenylalanine methyl ester hydrochloride, dl-p-chlorophenylalanine methyl ester hydrochloride, 4-chloro-dl-phe-ome hcl, methyl 2-amino-3-4-chlorophenyl propanoate, chloride, 4-chloro-dl-phe-ome hydrochloride, pcpa methyl ester hcl, 4-chloro-dl-phe-omecl
2733277
methyl 2-amino-3-(4-chlorophenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC=C(C=C1)Cl)N.Cl
This item is not returnable. View return policy
14173-40-1
C10H13Cl2NO2
250.119
MFCD00012530
GCBCWTWQAFLKJG-UHFFFAOYSA-N
4-chloro-dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, h-dl-phe 4-cl-ome.hcl, dl-4-chlorophenylalanine methyl ester hydrochloride, dl-p-chlorophenylalanine methyl ester hydrochloride, 4-chloro-dl-phe-ome hcl, methyl 2-amino-3-4-chlorophenyl propanoate, chloride, 4-chloro-dl-phe-ome hydrochloride, pcpa methyl ester hcl, 4-chloro-dl-phe-omecl
2733277
methyl 2-amino-3-(4-chlorophenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC=C(C=C1)Cl)N.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 186°C to 189°C (decomposition)
Formula Weight 250.12
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material 4-Chloro-DL-phenylalanine methyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'