missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%

Brand:  Alfa Aesar™ H63873.06

Code : NEW

Additional Details : CAS Number : 33965-47-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428566

  • £198.50 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-D-phenylalanine methyl ester hydrochloride
250.12
H-D-Phe(4-Cl)-Ome.HCl; (R)-Methyl 2-amino-3-(4-chlorophenyl)propionate hydrochloride
33965-47-8
MFCD00236920

Specifications

4-Chloro-D-phenylalanine methyl ester hydrochloride
Powder
250.12
H-D-Phe(4-Cl)-Ome.HCl; (R)-Methyl 2-amino-3-(4-chlorophenyl)propionate hydrochloride
5g
33965-47-8
98%
MFCD00236920
White
Videos
SDS