missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-methylphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7044359
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
938-95-4
C9H9ClO2
184.619
MFCD00044670
YOZILQVNIWNPFP-UHFFFAOYSA-N
2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
102525
2-(4-chlorophenyl)propanoic acid
CC(C1=CC=C(C=C1)Cl)C(=O)O
This item is not returnable. View return policy
938-95-4
C9H9ClO2
184.619
MFCD00044670
YOZILQVNIWNPFP-UHFFFAOYSA-N
2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
102525
2-(4-chlorophenyl)propanoic acid
CC(C1=CC=C(C=C1)Cl)C(=O)O

4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 55°C to 59°C
Boiling Point 121°C to 126°C (0.2 mmHg)
Quantity 5 g
Beilstein 2328710
Solubility Information Soluble in water.
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 4-Chloro-alpha-methylphenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'