missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-6-methylpyrimidine, 97%

Brand:  Alfa Aesar™ H64667.MD

Code : NEW

Additional Details : CAS Number : 3435-25-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15441969

  • £20.30 / 250mg


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4-Chloro-6-methylpyrimidine
128.56
108656
3435-25-4
MFCD02322991

Specifications

4-Chloro-6-methylpyrimidine
Solid
Pale yellow
128.56
108656
3435-25-4
97%
250mg
MFCD02322991
Videos
SDS