missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-trifluoromethyl-o-phenylenediamine, 95%, Thermo Scientific™

Catalog Number p-7052450
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
157590-59-5
C7H6ClF3N2
210.584
MFCD00119550
BUSGYQISMPLVIJ-UHFFFAOYSA-N
4-chloro-5-trifluoromethyl benzene-1,2-diamine, 2-amino-4-chloro-5-trifluoromethyl aniline, 4-chloro-5-trifluoromethyl-o-phenylenediamine, 2-amino-4-chloro-5-trifluoromethyl phenylamine, 2-chloro-4,5-diaminobenzotrifluoride, 5-chloro-4-trifluoromethyl benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-trifluoromethyl, aminochlorotrifluoromethylphenylamine, 3,4-diamino-6-chlorobenzotrifluoride
2773497
4-chloro-5-(trifluoromethyl)benzene-1,2-diamine
C1=C(C(=CC(=C1N)N)Cl)C(F)(F)F
This item is not returnable. View return policy
157590-59-5
C7H6ClF3N2
210.584
MFCD00119550
BUSGYQISMPLVIJ-UHFFFAOYSA-N
4-chloro-5-trifluoromethyl benzene-1,2-diamine, 2-amino-4-chloro-5-trifluoromethyl aniline, 4-chloro-5-trifluoromethyl-o-phenylenediamine, 2-amino-4-chloro-5-trifluoromethyl phenylamine, 2-chloro-4,5-diaminobenzotrifluoride, 5-chloro-4-trifluoromethyl benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-trifluoromethyl, aminochlorotrifluoromethylphenylamine, 3,4-diamino-6-chlorobenzotrifluoride
2773497
4-chloro-5-(trifluoromethyl)benzene-1,2-diamine
C1=C(C(=CC(=C1N)N)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-5-trifluoromethyl-o-phenylenediamine has application in dietary supplements, pharmaceutical or cosmeticeuticals.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 61°C to 63°C
Color White to Brown
Quantity 250 mg
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 210.59
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Chloro-5-trifluoromethyl-o-phenylenediamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'