missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog Number 11985261
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
67073-39-6
C6H6ClN3O2
187.58
MFCD03427594
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
5702618
4-chloro-5-nitrobenzene-1,2-diamine
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
This item is not returnable. View return policy
67073-39-6
C6H6ClN3O2
187.58
MFCD03427594
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
5702618
4-chloro-5-nitrobenzene-1,2-diamine
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 189.0°C to 193.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 187.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-chloro-5-nitro-o-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-5-nitro-o-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'