missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-methylbenzene-1,2-diamine, ≥95%, Thermo Scientific™

Catalog Number p-2701691
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
63155-04-4
C7H9ClN2
156.613
MFCD00221471
HOFKXNBVTNUDSH-UHFFFAOYSA-N
2801468
4-chloro-5-methylbenzene-1,2-diamine
CC1=CC(=C(C=C1Cl)N)N
This item is not returnable. View return policy
63155-04-4
C7H9ClN2
156.613
MFCD00221471
HOFKXNBVTNUDSH-UHFFFAOYSA-N
2801468
4-chloro-5-methylbenzene-1,2-diamine
CC1=CC(=C(C=C1Cl)N)N
CAS Min % 95.0
Assay Percent Range 95%
Synonym 1,2-benzenediamine,4-chloro-5-methyl, 4-chloro-5-methyl-1,2-phenylenediamine, 4mrh, pubchem22761, 2-chloro-4,5-diaminotoluene, 2-amino-4-chloro-5-methylphenyl amine, 4-chloro-5-methyl-benzene-1,2-diamine, 5-chloro-4-methyl-1,2-phenylenediamine, 1,2-benzenediamine, 4-chloro-5-methyl, 4-chloranyl-5-methyl-benzene-1,2-diamine
Formula Weight 156.62
Percent Purity 95%
Quantity 1 g
Packaging Amber glass bottle
Chemical Name or Material 4-Chloro-5-methylbenzene-1, 2-diamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'