missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Maybridge™

£38.44 - £214.03

Chemical Identifiers

CAS 189681-04-7
Molecular Formula C10H5ClN2S2
Molecular Weight (g/mol) 252.734
MDL Number MFCD00174019
InChI Key VNWBUFWVNCUUKY-UHFFFAOYSA-N
Synonym 4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
PubChem CID 735732
IUPAC Name 4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
SMILES C1=CSC(=C1)C2=CSC3=C2C(=NC=N3)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10028961
SDS View Documents
Maybridge
AW00007DA
1g
£38.44
1g
View alternative products
Save Up to 
»
Estimated Shipment
07-12-2020
Please to purchase this item. Need a web account? Register with us today!
10284874
SDS View Documents
Maybridge
AW00007EA
10g
£214.03
10g
View alternative products
Save Up to 
»
Estimated Shipment
07-12-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

189681-04-7
252.734
VNWBUFWVNCUUKY-UHFFFAOYSA-N
735732
C1=CSC(=C1)C2=CSC3=C2C(=NC=N3)Cl
C10H5ClN2S2
MFCD00174019
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine

Specifications

4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine
1g
94.5
95%
MFCD00174019
VNWBUFWVNCUUKY-UHFFFAOYSA-N
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
735732
95%
Amber Glass Bottle
189681-04-7
100.0
C10H5ClN2S2
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
C1=CSC(=C1)C2=CSC3=C2C(=NC=N3)Cl
252.734
252.75
SDS