missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Thermo Scientific™

Catalog Number 10284874
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
189681-04-7
C10H5ClN2S2
252.73
MFCD00174019
VNWBUFWVNCUUKY-UHFFFAOYSA-N
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
735732
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
This item is not returnable. View return policy
189681-04-7
C10H5ClN2S2
252.73
MFCD00174019
VNWBUFWVNCUUKY-UHFFFAOYSA-N
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
735732
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
IUPAC Name 4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
Formula Weight 252.75
Percent Purity 95%
Chemical Name or Material 4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'