missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-sulfamoyl-N-[2-(trifluoromethyl)phenyl]benzamide, 95%

Brand:  Alfa Aesar™ H65189.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449446

  • £137.80 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-3-sulfamoyl-N-[2-(trifluoromethyl)phenyl]benzamide
378.75
MFCD27975732

Specifications

4-Chloro-3-sulfamoyl-N-[2-(trifluoromethyl)phenyl]benzamide
Solid
378.75
1g
95%
MFCD27975732
Videos
SDS