missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzotrifluoride, 98%, Thermo Scientific™

Catalog Number p-89821
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
121-17-5
C7H3ClF3NO2
225.55
MFCD00007084
TZGFQIXRVUHDLE-UHFFFAOYSA-N
4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
8462
1-chloro-2-nitro-4-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
This item is not returnable. View return policy
121-17-5
C7H3ClF3NO2
225.55
MFCD00007084
TZGFQIXRVUHDLE-UHFFFAOYSA-N
4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
8462
1-chloro-2-nitro-4-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.0°C
Color Yellow
Density 1.5110g/mL
Boiling Point 222.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(NO2)CF3
Refractive Index 1.4883 to 1.4903
Quantity 10 g
Specific Gravity 1.511
Solubility Information Solubility in water: 660mg/l water (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 225.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'