missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-nitroanisole, 98%

Brand:  Alfa Aesar™ A12282.14

Code : NEW

Additional Details : CAS Number : 10298-80-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11465003

  • £25.40 / 25g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-3-nitroanisole
C7H6ClNO3
MFCD00007077
1-Chloro-4-methoxy-2-nitrobenzene; 4-Methoxy-2-nitrochlorobenzene
10298-80-3
187.58
640872

Specifications

4-Chloro-3-nitroanisole
42° to 45°C
10298-80-3
C7H6ClNO3
MFCD00007077
1-Chloro-4-methoxy-2-nitrobenzene; 4-Methoxy-2-nitrochlorobenzene
>110°C (230°F)
25g
98%
187.58
640872
Videos
SDS