missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number p-7029154
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
883500-51-4
C8H6ClFO2
188.582
MFCD04115861
QIYNMJFDEWFEKJ-UHFFFAOYSA-N
4-chloro-3-fluorophenylacetic acid, 2-4-chloro-3-fluorophenyl acetic acid, 4-chloro-3-fluorophenylaceticacid, 4-chloro-3-fluorophenyl acetic acid, pubchem4301, acmc-209qsi, 2-4-chloro-3-fluoro-phenyl acetic acid, 4-chloro-3-fluorphenylaceticacid, 4-chloro-3-fluoro-phenyl-acetic acid
3814954
2-(4-chloro-3-fluorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)Cl
This item is not returnable. View return policy
883500-51-4
C8H6ClFO2
188.582
MFCD04115861
QIYNMJFDEWFEKJ-UHFFFAOYSA-N
4-chloro-3-fluorophenylacetic acid, 2-4-chloro-3-fluorophenyl acetic acid, 4-chloro-3-fluorophenylaceticacid, 4-chloro-3-fluorophenyl acetic acid, pubchem4301, acmc-209qsi, 2-4-chloro-3-fluoro-phenyl acetic acid, 4-chloro-3-fluorphenylaceticacid, 4-chloro-3-fluoro-phenyl-acetic acid
3814954
2-(4-chloro-3-fluorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 88°C to 91°C
Quantity 1 g
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'