missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorophenol, 98%, Thermo Scientific™

Catalog Number p-87573
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
348-60-7
C6H4ClFO
146.55
MFCD00042583
XLHYAEBESNFTCA-UHFFFAOYSA-N
3-fluoro-4-chlorophenol, 4-chloro-3-fluoro-phenol, phenol, 4-chloro-3-fluoro, pubchem1493, acmc-1ctvv, 3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluorophenol;
2724523
4-chloro-3-fluorophenol
OC1=CC=C(Cl)C(F)=C1
This item is not returnable. View return policy
348-60-7
C6H4ClFO
146.55
MFCD00042583
XLHYAEBESNFTCA-UHFFFAOYSA-N
3-fluoro-4-chlorophenol, 4-chloro-3-fluoro-phenol, phenol, 4-chloro-3-fluoro, pubchem1493, acmc-1ctvv, 3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluorophenol;
2724523
4-chloro-3-fluorophenol
OC1=CC=C(Cl)C(F)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color Colorless to Orange
Boiling Point 102.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula OHC6H3ClF
Quantity 1 g
Formula Weight 146.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-fluorophenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-fluorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'