missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-fluorobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H64411.03

Code : NEW

Additional Details : CAS Number : 32137-20-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491609

  • £12.51 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-3-fluorobenzotrifluoride
198.54
2093904
32137-20-5
MFCD00060646
1-Chloro-2-fluoro-4-(trifluoromethyl)benzene

Specifications

4-Chloro-3-fluorobenzotrifluoride
135°C
1g
1.4335
98%
MFCD00060646
1-Chloro-2-fluoro-4-(trifluoromethyl)benzene
1.49g/mL
Pale yellow
32137-20-5
Liquid
198.54
2093904
Videos
SDS