missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15436987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323955-63-0
C10H10ClFO3
232.635
MFCD19687188
JLMJTUGWXPDCKF-UHFFFAOYSA-N
4-chloro-3-ethoxy-2-fluorophenylacetic acid, 4-chloro-3-ethoxy-2-fluorophenyl acetic acid, 2-4-chloro-3-ethoxy-2-fluorophenyl acetic acid
66523623
2-(4-chloro-3-ethoxy-2-fluorophenyl)acetic acid
CCOC1=C(C=CC(=C1F)CC(=O)O)Cl
This item is not returnable. View return policy
1323955-63-0
C10H10ClFO3
232.635
MFCD19687188
JLMJTUGWXPDCKF-UHFFFAOYSA-N
4-chloro-3-ethoxy-2-fluorophenylacetic acid, 4-chloro-3-ethoxy-2-fluorophenyl acetic acid, 2-4-chloro-3-ethoxy-2-fluorophenyl acetic acid
66523623
2-(4-chloro-3-ethoxy-2-fluorophenyl)acetic acid
CCOC1=C(C=CC(=C1F)CC(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 232.64
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material 4-Chloro-3-ethoxy-2-fluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'