missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number 15416757
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1323966-33-1
C9H8ClFO3
218.61
MFCD19687215
AMAJHTMBCOHVIL-UHFFFAOYSA-N
66523585
4-chloro-3-ethoxy-2-fluorobenzoic acid
CCOC1=C(Cl)C=CC(C(O)=O)=C1F
This item is not returnable. View return policy
1323966-33-1
C9H8ClFO3
218.61
MFCD19687215
AMAJHTMBCOHVIL-UHFFFAOYSA-N
66523585
4-chloro-3-ethoxy-2-fluorobenzoic acid
CCOC1=C(Cl)C=CC(C(O)=O)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 218.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-3-ethoxy-2-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'