missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7052489
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-26-2
C9H9ClFNO2
217.624
MFCD19687217
GZFQGYBTUBVMAM-UHFFFAOYSA-N
66523587
4-chloro-3-ethoxy-2-fluorobenzamide
CCOC1=C(C=CC(=C1F)C(=O)N)Cl
This item is not returnable. View return policy
1323966-26-2
C9H9ClFNO2
217.624
MFCD19687217
GZFQGYBTUBVMAM-UHFFFAOYSA-N
66523587
4-chloro-3-ethoxy-2-fluorobenzamide
CCOC1=C(C=CC(=C1F)C(=O)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 217.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-3-ethoxy-2-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'