missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)quinoline, 98%, Thermo Scientific™

Catalog Number p-7081036
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1701-24-2
C10H5ClF3N
231.602
MFCD00153105
ONNDFDQMHCNEGF-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl quinoline, 2-trifluoromethyl-4-chloroquinoline, 4-chloro-2-trifluoromethyl-quinoline, quinoline, 4-chloro-2-trifluoromethyl, pubchem5854, buttpark 29\04-05, acmc-209e0j
2736709
4-chloro-2-(trifluoromethyl)quinoline
C1=CC=C2C(=C1)C(=CC(=N2)C(F)(F)F)Cl
This item is not returnable. View return policy
1701-24-2
C10H5ClF3N
231.602
MFCD00153105
ONNDFDQMHCNEGF-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl quinoline, 2-trifluoromethyl-4-chloroquinoline, 4-chloro-2-trifluoromethyl-quinoline, quinoline, 4-chloro-2-trifluoromethyl, pubchem5854, buttpark 29\04-05, acmc-209e0j
2736709
4-chloro-2-(trifluoromethyl)quinoline
C1=CC=C2C(=C1)C(=CC(=N2)C(F)(F)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 31°C to 33°C
Flash Point 107°C (225°F)
Quantity 1 g
Formula Weight 231.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-2- (trifluoromethyl)quinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'