missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Catalog Number p-2701655
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175205-77-3
C7H5ClF3NO
211.568
MFCD00153102
UCFTYLMKCJPWBX-UHFFFAOYSA-N
2781526
4-chloro-2-(trifluoromethoxy)aniline
C1=CC(=C(C=C1Cl)OC(F)(F)F)N
This item is not returnable. View return policy
175205-77-3
C7H5ClF3NO
211.568
MFCD00153102
UCFTYLMKCJPWBX-UHFFFAOYSA-N
2781526
4-chloro-2-(trifluoromethoxy)aniline
C1=CC(=C(C=C1Cl)OC(F)(F)F)N
CAS Min % 97.0
Assay Percent Range 97%
Synonym 4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine
Formula Weight 211.57
Percent Purity 97%
Quantity 250 mg
Packaging Amber Glass Bottle
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'