missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Synonym | 4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine |
Formula Weight | 211.57 |
Percent Purity | 97% |
Quantity | 250 mg |
Packaging | Amber Glass Bottle |
Chemical Name or Material | 4-Chloro-2-(trifluoromethoxy)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'