missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylthiopyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4813090
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
49844-90-8
C5H5ClN2S
160.62
MFCD00006083
DFOHHQRGDOQMKG-UHFFFAOYSA-N
4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
97113
4-chloro-2-methylsulfanylpyrimidine
CSC1=NC=CC(Cl)=N1
This item is not returnable. View return policy
49844-90-8
C5H5ClN2S
160.62
MFCD00006083
DFOHHQRGDOQMKG-UHFFFAOYSA-N
4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
97113
4-chloro-2-methylsulfanylpyrimidine
CSC1=NC=CC(Cl)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2.0°C
Density 1.3810g/mL
Boiling Point 139.0°C to 140.0°C (36.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6004
Quantity 10 g
Beilstein 23, 371
Specific Gravity 1.381
Formula Weight 160.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylthiopyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-2-methylthiopyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'