missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-methylquinoline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | ∼25°C |
Density | 0.881 |
Boiling Point | 269°C to 270°C |
Flash Point | >110°C (230°F) |
Molecular Formula | C10H8ClN |
Refractive Index | 1.622 |
Quantity | 1 g |
Beilstein | 3522 |
Synonym | 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl |
Solubility Information | Slightly soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'