missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylaniline, 98%, ACROS Organics™

Brand:  Acros Organics 109375000

Code : 29

Additional Details : CAS Number : 95-69-2 Weight : 0.55000kg Transport : UN number : 2239 Chem class : 1 Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10640873

  • £545.00 / 500g


Estimated Shipment
12-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Chloro-2-methylaniline
95-69-2
100.0
C7H8ClN
141.6
4-Chloro-o-toluidine
98%
97.5
98%
ClC6H3(CH3)NH2
MFCD00007842

Specifications

4-Chloro-2-methylaniline
124°C
29.0° to 30.0°C
98%
97.5
Glass bottle
98%
C7H8ClN
141.6
4-Chloro-o-toluidine
1.1900g/mL
241.0°C
500g
95-69-2
100.0
1.5845 to 1.5865
98%
ClC6H3(CH3)NH2
MFCD00007842
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Chloro-2-methylaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / A§
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

SDS