missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxy-5-methylaniline, 85%, Tech., Thermo Scientific™

Catalog Number p-88754
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
6376-14-3
C8H10ClNO
171.62
MFCD00007847
XBAPOWUMJRIKAV-UHFFFAOYSA-N
chlorocresidine, 4-chloro-5-methyl-o-anisidine, o-anisidine, 4-chloro-5-methyl, 4-chloro-2-methoxy-5-methylphenylamine, 4-chloro-2-methoxy-5-methylanilin, 2-methoxy-5-methyl-4-chloroaniline, 4-chloro-2-methoxy-5-methyl-aniline, 4-chloro-2-methoxy-5-methyl-phenylamine, benzenamine,4-chloro-2-methoxy-5-methyl, tech.,
80782
4-chloro-2-methoxy-5-methylaniline
COC1=CC(Cl)=C(C)C=C1N
This item is not returnable. View return policy
6376-14-3
C8H10ClNO
171.62
MFCD00007847
XBAPOWUMJRIKAV-UHFFFAOYSA-N
chlorocresidine, 4-chloro-5-methyl-o-anisidine, o-anisidine, 4-chloro-5-methyl, 4-chloro-2-methoxy-5-methylphenylamine, 4-chloro-2-methoxy-5-methylanilin, 2-methoxy-5-methyl-4-chloroaniline, 4-chloro-2-methoxy-5-methyl-aniline, 4-chloro-2-methoxy-5-methyl-phenylamine, benzenamine,4-chloro-2-methoxy-5-methyl, tech.,
80782
4-chloro-2-methoxy-5-methylaniline
COC1=CC(Cl)=C(C)C=C1N
CAS Min % 80.0
CAS Max % 100.0
Melting Point 97.0°C to 103.0°C
Color Black
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Quantity 5 g
Formula Weight 171.63
Percent Purity 85%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline, tech., 85%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'