missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-iodoanisole, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
An independent synthesis of 4-chloro-2-iodoanisole has shown that this is chlorinating agents, brominating agents and iodinating agents. 4-Chloro-2-iodoanisole (2) was demethylated and subsequently alkylated with 4-chloro-2-fluorobenzyl bromide, an aryl Grignard reagent was formed with it.
Solubility
Soluble in methanol.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
An independent synthesis of 4-chloro-2-iodoanisole has shown that this is chlorinating agents, brominating agents and iodinating agents. 4-Chloro-2-iodoanisole (2) was demethylated and subsequently alkylated with 4-chloro-2-fluorobenzyl bromide, an aryl Grignard reagent was formed with it.
Solubility
Soluble in methanol.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Melting Point | 44°C to 48°C |
Boiling Point | 133°C to 135°C (15 mmHg) |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 268.48 |
Percent Purity | 98% |
Chemical Name or Material | 4-Chloro-2-iodoanisole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'