missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoropyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15411839
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
34941-92-9
C5H3ClFN
131.534
MFCD04112503
GNJKJKBURMCLOR-UHFFFAOYSA-N
2-fluoro-4-chloropyridine, 4-chloro-2-fluoro-pyridine, pyridine, 4-chloro-2-fluoro, pubchem13535, pyridine,4-chloro-2-fluoro
2762839
4-chloro-2-fluoropyridine
C1=CN=C(C=C1Cl)F
This item is not returnable. View return policy
34941-92-9
C5H3ClFN
131.534
MFCD04112503
GNJKJKBURMCLOR-UHFFFAOYSA-N
2-fluoro-4-chloropyridine, 4-chloro-2-fluoro-pyridine, pyridine, 4-chloro-2-fluoro, pubchem13535, pyridine,4-chloro-2-fluoro
2762839
4-chloro-2-fluoropyridine
C1=CN=C(C=C1Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 1634727
Formula Weight 131.53
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'