missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2-fluoropyridine, 95%

Brand:  Alfa Aesar™ H64557.06

Code : NEW

Additional Details : CAS Number : 34941-92-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401839

  • £219.00 / 5g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

34941-92-9
131.534
GNJKJKBURMCLOR-UHFFFAOYSA-N
2762839
C1=CN=C(C=C1Cl)F
C5H3ClFN
MFCD04112503
2-fluoro-4-chloropyridine, 4-chloro-2-fluoro-pyridine, pyridine, 4-chloro-2-fluoro, pubchem13535, pyridine,4-chloro-2-fluoro
4-chloro-2-fluoropyridine

Specifications

4-Chloro-2-fluoropyridine
34941-92-9
MFCD04112503
2-fluoro-4-chloropyridine, 4-chloro-2-fluoro-pyridine, pyridine, 4-chloro-2-fluoro, pubchem13535, pyridine,4-chloro-2-fluoro
C1=CN=C(C=C1Cl)F
131.534
131.53
95%
5g
C5H3ClFN
1634727
GNJKJKBURMCLOR-UHFFFAOYSA-N
4-chloro-2-fluoropyridine
2762839
Solid
Videos
SDS Documents
Certificates