missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenol, +98%, ACROS Organics™

Brand:  Acros Organics 378210250

Code : 29

Additional Details : CAS Number : 348-62-9 Weight : 0.08000kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10140613

  • £167.80 / 25g


Estimated Shipment
22-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-2-fluorophenol
98.0
≥98.0%
C6H4ClFO
146.55
348-62-9
100.0
98+%
ClC6H3FOH
MFCD00075030

Specifications

4-Chloro-2-fluorophenol
Authentic
25g
98.0
100.0
≥98.0%
C6H4ClFO
146.55
1.3780g/mL
Colorless to pink
348-62-9
Glass bottle
Liquid or Low Melting Solid
98+%
ClC6H3FOH
MFCD00075030
Videos
SDS