missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenol, 98+%, Thermo Scientific™

Catalog Number p-3212037
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
181975
4-chloro-2-fluorophenol
OC1=CC=C(Cl)C=C1F
This item is not returnable. View return policy
348-62-9
C6H4ClFO
146.55
MFCD00075030
ZKMUKBBWORLNLA-UHFFFAOYSA-N
2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
181975
4-chloro-2-fluorophenol
OC1=CC=C(Cl)C=C1F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 20°C
Color Colorless to Pink
Density 1.3780g/mL
Boiling Point 103°C to 104°C (50.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3FOH
Refractive Index 1.5330 to 1.535
Quantity 5 g
Specific Gravity 1.378
Solubility Information Solubility in water: insoluble
Formula Weight 146.55
Percent Purity 98+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Chloro-2-fluorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'