missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65522.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469706

  • £99.64 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-sulfamoylbenzamide
346.74
MFCD28098801

Specifications

4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-sulfamoylbenzamide
Solid
346.74
1g
95%
MFCD28098801
Videos
SDS