missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2-fluoro-5-sulfamoyl-N-[3-(trifluoromethyl)phenyl]benzamide, 95%

Brand:  Alfa Aesar™ H65828.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489926

  • £79.71 / 1g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-2-fluoro-5-sulfamoyl-N-[3-(trifluoromethyl)phenyl]benzamide
396.74
MFCD28098829

Specifications

4-Chloro-2-fluoro-5-sulfamoyl-N-[3-(trifluoromethyl)phenyl]benzamide
Solid
396.74
1g
95%
MFCD28098829
Videos
SDS