missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific™

Catalog Number p-7058174
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
35112-05-1
C7H3ClFNO4
219.552
MFCD04115725
KAONECPATVTFST-UHFFFAOYSA-N
4-chloro-2-fluoro-5-nitrobenzoic acid, 4-chloro-2-fluoro-5-nitrobenzoicacid, 4-chloro-2-fluoro-5-nitro-benzoic acid, 2-fluoro-4-chloro-5-nitrobenzoic acid, benzoic acid, 4-chloro-2-fluoro-5-nitro, pubchem22286, acmc-1ah9o, ksc573m6p, 5-carboxy-2-chloro-4-fluoronitrobenzene
3774553
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)F)C(=O)O
This item is not returnable. View return policy
35112-05-1
C7H3ClFNO4
219.552
MFCD04115725
KAONECPATVTFST-UHFFFAOYSA-N
4-chloro-2-fluoro-5-nitrobenzoic acid, 4-chloro-2-fluoro-5-nitrobenzoicacid, 4-chloro-2-fluoro-5-nitro-benzoic acid, 2-fluoro-4-chloro-5-nitrobenzoic acid, benzoic acid, 4-chloro-2-fluoro-5-nitro, pubchem22286, acmc-1ah9o, ksc573m6p, 5-carboxy-2-chloro-4-fluoronitrobenzene
3774553
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 219.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-5-nitrobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'