missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-3-methoxybenzyl bromide, 97%, Thermo Scientific™

Catalog Number 15418305
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-35-3
C8H7BrClFO
253.50
MFCD19687182
PSWKNVCWOPRAMU-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzyl bromide, 1-bromomethyl-4-chloro-2-fluoro-3-methoxybenzene
66523607
1-(bromomethyl)-4-chloro-2-fluoro-3-methoxybenzene
COC1=C(Cl)C=CC(CBr)=C1F
This item is not returnable. View return policy
1323966-35-3
C8H7BrClFO
253.50
MFCD19687182
PSWKNVCWOPRAMU-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzyl bromide, 1-bromomethyl-4-chloro-2-fluoro-3-methoxybenzene
66523607
1-(bromomethyl)-4-chloro-2-fluoro-3-methoxybenzene
COC1=C(Cl)C=CC(CBr)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3261
Formula Weight 253.5
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluoro-3-methoxybenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'