missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-3-methoxybenzyl alcohol, 97%, Thermo Scientific™

Catalog Number 15428675
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-21-7
C8H8ClFO2
190.598
MFCD19687181
ULQDMTWMSOVQRT-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzyl alcohol, 4-chloro-2-fluoro-3-methoxyphenyl methanol
66523606
(4-chloro-2-fluoro-3-methoxyphenyl)methanol
COC1=C(C=CC(=C1F)CO)Cl
This item is not returnable. View return policy
1323966-21-7
C8H8ClFO2
190.598
MFCD19687181
ULQDMTWMSOVQRT-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxybenzyl alcohol, 4-chloro-2-fluoro-3-methoxyphenyl methanol
66523606
(4-chloro-2-fluoro-3-methoxyphenyl)methanol
COC1=C(C=CC(=C1F)CO)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 190.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluoro-3-methoxybenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'