missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2-fluoro-3-methoxybenzoic acid, 97%

Brand:  Alfa Aesar™ H33114.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15415907

  • £151.90 / 5g


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-2-fluoro-3-methoxybenzoic acid
204.59
MFCD19687204

Specifications

4-Chloro-2-fluoro-3-methoxybenzoic acid
Solid
204.59
5g
97%
MFCD19687204
Videos
SDS