missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-3-methoxyaniline, 97%, Thermo Scientific™

Catalog Number 15427197
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-39-7
C7H7ClFNO
175.587
MFCD19687207
ICWULCQRXSKPBC-UHFFFAOYSA-N
17856680
4-chloro-2-fluoro-3-methoxyaniline
COC1=C(C=CC(=C1F)N)Cl
This item is not returnable. View return policy
1323966-39-7
C7H7ClFNO
175.587
MFCD19687207
ICWULCQRXSKPBC-UHFFFAOYSA-N
17856680
4-chloro-2-fluoro-3-methoxyaniline
COC1=C(C=CC(=C1F)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2811
Formula Weight 175.59
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluoro-3-methoxyaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'