4-Chloro-2,8-bis(trifluoromethyl)quinoline, 97%, ACROS Organics™


Overview
£10.40 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 306022500

250MG 4-Chloro-2,8-bis(trifluoromethyl)quinoline,97%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 83012-13-9 Weight: 0.05025kg


Quantity 250mg

Product Code. 10063740

Quantity Price
1 £ 26.0 / 250mg
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Name Note 97%
CAS 83012-13-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H4ClF6N
Formula Weight 299.6
MDL Number MFCD00075104

Specification

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 299.6
Quantity 250mg
Color Beige
CAS Min % 96.0
EINECS Number 280-132-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 83012-13-9
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00075104
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Name Note 97%
Molecular Formula C11H4ClF6N
CAS Max % 100.0
Chemical Name or Material 4-Chloro-2, 8-bis(trifluoromethyl)quinoline
Melting Point 45° to 49°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.