missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2,5-difluorobenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 153°C to 158°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | ClC6H2F2(COOH) |
Quantity | 10 g |
Formula Weight | 192.55 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'