missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-86795
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
132794-07-1
C7H2ClF2O2
191.54
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-M
2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
2733274
4-chloro-2,5-difluorobenzoic acid
[O-]C(=O)C1=CC(F)=C(Cl)C=C1F
This item is not returnable. View return policy
132794-07-1
C7H2ClF2O2
191.54
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-M
2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
2733274
4-chloro-2,5-difluorobenzoic acid
[O-]C(=O)C1=CC(F)=C(Cl)C=C1F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 153°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H2F2(COOH)
Quantity 1 g
Formula Weight 192.55
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2, 5-difluorobenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'