missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-2,5-difluorobenzaldehyde, 98%

Brand:  Alfa Aesar™ H64112.03

Code : NEW

Additional Details : CAS Number : 879093-02-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15411169

  • £23.30 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-2,5-difluorobenzaldehyde
176.55
14516595
879093-02-4
MFCD07782051

Specifications

4-Chloro-2,5-difluorobenzaldehyde
1g
Solid
176.55
14516595
Off-white
879093-02-4
98%
MFCD07782051
Videos
SDS