missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-1,2-dihydrophthalazin-1-one, 97%, Thermo Scientific™

Catalog Number p-2701640
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
2257-69-4
C8H5ClN2O
180.59
MFCD00102192
QCKGMJDOJRNSMS-UHFFFAOYSA-N
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
2775142
4-chloro-2H-phthalazin-1-one
ClC1=NNC(=O)C2=CC=CC=C12
This item is not returnable. View return policy
2257-69-4
C8H5ClN2O
180.59
MFCD00102192
QCKGMJDOJRNSMS-UHFFFAOYSA-N
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
2775142
4-chloro-2H-phthalazin-1-one
ClC1=NNC(=O)C2=CC=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.59
Percent Purity 97%
Chemical Name or Material 4-chloro-1, 2-dihydrophthalazin-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'