missing translation for 'onlineSavingsMsg'
Learn More

4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444670050

Code : 29

Additional Details : CAS Number : 797755-07-8 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11956061

  • £551.80 / 5g


Estimated Shipment
15-03-2018
Add to basket


Specifications

Chemical Identifiers

4-(Carboxymethyl)phenylboronic acid pinacol ester
C14H19BO4
2-(4-(4, 4, 5
97%
262.11

Specifications

4-(Carboxymethyl)phenylboronic acid pinacol ester
162° to 168°C
797755-07-8
Crystalline Powder or Powder
C14H19BO4
2-(4-(4, 4, 5
White to yellow
5g
96 % min. Consistent with structure (1H-NMR)
97%
262.11
Videos
SDS