missing translation for 'onlineSavingsMsg'
Learn More

4-Butylaniline, 97%, Thermo Scientific Chemicals

Catalog Number 11935241
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104-13-2
C10H15N
149.23
OGIQUQKNJJTLSZ-UHFFFAOYSA-N
4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine
7694
4-butylaniline
CCCCC1=CC=C(C=C1)N
This item is not returnable. View return policy
104-13-2
C10H15N
149.23
OGIQUQKNJJTLSZ-UHFFFAOYSA-N
4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine
7694
4-butylaniline
CCCCC1=CC=C(C=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.9450g/mL
Boiling Point 133.0°C to 134.0°C (14.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3C6H4NH2
Refractive Index 1.535
Quantity 100 g
Beilstein 12,I,503
Specific Gravity 0.945
Formula Weight 149.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Butylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Butylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'