missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzaldehyde, 99%, Thermo Scientific™

Catalog Number 10608824
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5736-88-9
C11H14O2
178.23
MFCD00003389
XHWMNHADTZZHGI-UHFFFAOYSA-N
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
79813
4-butoxybenzaldehyde
CCCCOC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
5736-88-9
C11H14O2
178.23
MFCD00003389
XHWMNHADTZZHGI-UHFFFAOYSA-N
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
79813
4-butoxybenzaldehyde
CCCCOC1=CC=C(C=C1)C=O
CAS Min % 98.5
CAS Max % 100.0
Color Red to Yellow
Boiling Point 148°C to 149°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)3OC6H4CHO
Refractive Index 1.5390 to 1.541
Quantity 50 g
Beilstein 08, II, 67
Formula Weight 178.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Butoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'