missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoveratrole 97%, ACROS Organics™

Brand:  Acros Organics 107540250

Code : 29

Additional Details : CAS Number : 2859-78-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10489010

  • £39.40 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

4-Bromoveratrole
2859-78-1
100.0
97%
BrC6H3(OCH3)2
MFCD00008381
97%
96.0
≥96.0%
C8H9BrO2
217.06

Specifications

4-Bromoveratrole
2859-78-1
100.0
Glass bottle
Liquid
97%
BrC6H3(OCH3)2
MFCD00008381
25g
97%
96.0
Authentic
1.5720 to 1.5740 (20°C, 589nm)
≥96.0%
C8H9BrO2
217.06
Clear colorless to slightly yellow
Videos
SDS