missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoveratrole, 97%, Thermo Scientific Chemicals

Catalog Number p-77682
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2859-78-1
C8H9BrO2
217.06
MFCD00008381
KBTMGSMZIKLAHN-UHFFFAOYSA-N
4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
76114
4-bromo-1,2-dimethoxybenzene
COC1=C(C=C(C=C1)Br)OC
This item is not returnable. View return policy
2859-78-1
C8H9BrO2
217.06
MFCD00008381
KBTMGSMZIKLAHN-UHFFFAOYSA-N
4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
76114
4-bromo-1,2-dimethoxybenzene
COC1=C(C=C(C=C1)Br)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5000g/mL
Boiling Point 255°C to 256°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OCH3)2
Refractive Index 1.5720 to 1.574
Quantity 5 g
Specific Gravity 1.5
Formula Weight 217.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromoveratrole, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoveratrole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'