missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoveratrole, 97%, Thermo Scientific Chemicals

Catalog Number p-7081238
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2859-78-1
C8H9BrO2
217.062
MFCD00008381
KBTMGSMZIKLAHN-UHFFFAOYSA-N
4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
76114
4-bromo-1,2-dimethoxybenzene
COC1=C(C=C(C=C1)Br)OC
This item is not returnable. View return policy
2859-78-1
C8H9BrO2
217.062
MFCD00008381
KBTMGSMZIKLAHN-UHFFFAOYSA-N
4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
76114
4-bromo-1,2-dimethoxybenzene
COC1=C(C=C(C=C1)Br)OC

4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

Solubility
Soluble in benzene, ether, toluene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.51
Boiling Point 255°C to 256°C
Flash Point 109°C (228°F)
Refractive Index 1.573
Quantity 25 g
Beilstein 2046884
Solubility Information Soluble in benzene,ether,toluene.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromoveratrole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'