missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene, Stabilized 96%, ACROS Organics™

Brand:  Acros Organics 154281000

Code : 29

Additional Details : CAS Number : 2039-82-9 Weight : 0.19000kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 11924521

  • £369.50 / 100mL


Estimated Shipment
12-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Bromostyrene
96%
H2C=CHC6H4Br
96%
C8H7Br
183.05

Specifications

4-Bromostyrene
Authentic
1.5930 to 1.5950
1.4000g/mL
88.0° to 89.0°C (16.0 mmHg)
75°C
96%
96%
H2C=CHC6H4Br
2039-82-9
Glass bottle
100 to 500ppm 4-tert.-butylcatechol
4.5°C
100mL
Liquid
Clear colorless to yellow
C8H7Br
183.05
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromostyrene, stabilized

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS