missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 15284754
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3964-04-3
208.06
SUXIPCHEUMEUSV-UHFFFAOYSA-N
CHEBI:46230
This item is not returnable. View return policy
3964-04-3
208.06
SUXIPCHEUMEUSV-UHFFFAOYSA-N
CHEBI:46230

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 32°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Molecular Formula C9H6BrN
Quantity 5 g
Synonym thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 4-thiazolecarboxylicacid, 4-carboxythiazole, 4-carboxy-1,3-thiazole, thiazole 4-carboxylic acid, acmc-2097eb, thiazol-4-carboxylic, ksc225i1n
SMILES BrC1=C2C=CC=CC2=NC=C1
IUPAC Name 1,3-thiazole-4-carboxylic acid
PubChem CID 304271
Formula Weight 208.06
Percent Purity 95%
Chemical Name or Material 4-Bromoquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoquinoline, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'